Tetrahydrofuran, extra dry 99.85% unstabilised, AcroSeal®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Butylene oxide, THF, Diethylene oxide

457070010 457071000 457070025
457070010.EA 84.3 GBP
457070010. 457071000. 457070025.
Tetrahydrofuran, extra dry 99.85% unstabilised, AcroSeal®
Tetrahydrofuran
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66 °C (760 mmHg)
Melting Pt: –108.4 °C
Density: 0.88 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
UN: 2056
ADR: 3 II
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.005 %
GC ≥99.80 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Acidity ≤0.0005 meq/g
Peroxides ≤0.0075 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR