VWR® ARISTAR®, Gold Standard Solutions for ICP-MS
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
456742MEA 573 GBP
456742M 85550.180
VWR® ARISTAR®, Gold Standard Solutions for ICP-MS
Standards Cationic Standards Gold Standards
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0,5% (typical)
  • Delivered with complete certificate of analysis
Certifications: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities