Magnesium oxide, tray, rectangular, Length: 100mm, Width: 25mm, Height: 12.7mm

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Magnesia usta

45634.KS
45634.KSEA 143 GBP
45634.KS
Magnesium oxide, tray, rectangular, Length: 100mm, Width: 25mm, Height: 12.7mm
Magnesium oxide
Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600
Melting Pt: 2800
Density: 3.58 g/cm³ (25 °C)
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR