VWR® ARISTAR®, Magnesium Standard Solution, 10000 mg/L Mg in dil. Nitric Acid (from MgO), Standard for ICP
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
455632DEA 257 GBP
455632D 455634F
VWR® ARISTAR®, Magnesium Standard Solution, 10000 mg/L Mg in dil. Nitric Acid (from MgO), Standard for ICP
Standards Cationic Standards Magnesium Standards
MgO in HNO₃ 2 to 5%.
Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery information: Supplied with certificate of analysis.