Calcium standard solution, 1.000 mg/l Ca in dil. nitric acid (from CaCO3) ARISTAR® standard for ICP
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
455162REA 199 GBP
455162R 455164T
Calcium standard solution, 1.000 mg/l Ca in dil. nitric acid (from CaCO3) ARISTAR® standard for ICP
Standards Cationic Standards Calcium Standards
CaCO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with Certificate of Analysis.