VWR® ARISTAR®, Cadmium Standard Solution, 10000 mg/L Cd in dil. Nitric Acid (from Cd), Standard for ICP
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
455154REA 483 GBP
455154R 455152P
VWR® ARISTAR®, Cadmium Standard Solution, 10000 mg/L Cd in dil. Nitric Acid (from Cd), Standard for ICP
Standards Cationic Standards Cadmium Standards
Cd in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.