High temperature tube furnaces with Tmax 1600 °C, RHTC series
Supplier: NABERTHERM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
RC183WAN RC283WAN RC383WAN
455-2509EA 8410 GBP
455-2509 455-2510 455-2511
High temperature tube furnaces with Tmax 1600 °C, RHTC series
Ovens Tube Furnaces
These compact tube furnaces with a SiC rod heating and integrated switchgear with controller can be used universally for many processes. They represent an inexpensive variant in the high-temperature range. The standard mounting options for accessories make them flexible for use in a wide range of applications.
  • The SiC heating elements are arranged parallel to the working tube providing excellent temperature uniformity
  • Equipped with professional P580 controller with touch screen display, featuring 50 freely adjustable programs and 40 segments and 2 to 6
More Product Information