Palladium 5% on activated carbon paste (50% water) A405032-5

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

45499.06 45499.14 45499.22
45499.06EA 75.2 GBP
45499.06 45499.14 45499.22
Palladium 5% on activated carbon paste (50% water) A405032-5
Palladium

CAS No.: 7440-05-3 Appearance: Black Odour: Odourless MDL No.: MFCD03457879

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR