Zinc oxide substrate 10x10x0.5mm, polished one side 0001 orientation

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Zinci oxidum, Zinc (II) oxide

45486.KT 45486.KU
45486.KTEA 680 GBP
45486.KT 45486.KU
Zinc oxide substrate 10x10x0.5mm, polished one side 0001 orientation
Zinc oxide

CAS No.: 1314-13-2 Molecular Formula: ZnO Formula Weight: 81.37 Odour: Odourless MDL No.: MFCD00011109

Formula: ZnO
Melting Pt: 1975 °C
Density: 5.61 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-171-9
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 13,10200

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR