Bismuth(III) sulphide ≥99.999% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

45463.30 45463.18
45463.30EA 1093 GBP
45463.30 45463.18
Bismuth(III) sulphide ≥99.999% (metals basis)
Bismuth (III) sulphide

Purity: 99.999% CAS No.: 1345-07-9 Molecular Formula: Bi2S3 Formula Weight: 514.15 Physical Form: Lump Density: 7.7 Appearance: Gray MDL No.: MFCD00014204

Formula: Bi₂S₃
Melting Pt: 763 °C
Density: 7.7
Storage Temperature: Ambient
MDL Number: MFCD00014204
CAS Number: 1345-07-9
EINECS: 215-716-0
Merck Index: 12,01329

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR