D-Glucose dehydrogenase

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

45402.4I
45402.4IEA 95.9 GBP
45402.4I
D-Glucose dehydrogenase
Enzymes

Applications: Diagnostics and cofactor regeneration of NAD(P)H in reductive processes.


Physical Form: Powder Appearance: Pale yellow MDL No.: MFCD00131181


Applications: Diagnostics and cofactor regeneration of NAD(P)H in reductive processes

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR