Optical windows
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
45387.KS 45386.KS 45380.KS 45560.KS 45666.KS 45813.KS 45856.KS 45925.KS 45986.KS 46057.KS 46089.KS
45387.KSEA 227 GBP
45387.KS 45386.KS 45380.KS 45560.KS 45666.KS 45813.KS 45856.KS 45925.KS 45986.KS 46057.KS 46089.KS
Optical windows
Optical Windows
These optical windows are ideally stored at ambient temperatures.
  • Clear aperture 85%
  • Surface finish 60/40 or 40/20