2,4,6-Tris(dimethylaminomethyl)phenol, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

45348-250ML-F
45348-250ML-FEA 77.8 GBP
45348-250ML-F
2,4,6-Tris(dimethylaminomethyl)phenol, Sigma-Aldrich®
2,4,6-Tris(dimethylaminomethyl)phenol
Formula: C₁₅H₂₇N₃O
Boiling Pt: 130…135 °C (1 mmHg)
Density: 0.974 g/cm³ (15 °C)
MDL Number: MFCD00008330
CAS Number: 90-72-2
EINECS: 202-013-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65518c3f6cd389d27c61916081b99816fdb6200b&vid=65518c3f6cd389d27c61916081b99816fdb6200b&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR