Rhodium standard solution, 10 mg/L Rh in 20% hydrochloric acid (from RhCl₃), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

45320.AEEA 163 GBP
45320.AE
Rhodium standard solution, 10 mg/L Rh in 20% hydrochloric acid (from RhCl₃), standard for ICP, Specpure®
Standards Cationic Standards Rhodium Standards

Molecular Formula: RhCl₃ in 2% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011201


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR