Sulphur in heavy mineral oil standard solutions, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Sulphur in heavy mineral oil standard solutions, Specpure®
Standards Petroleum Industry Standards

Specpure® calibration standards, designed specifically for the analysis of sulphur in petroleum products by X-ray fluorescence spectrometry.


Physical Form: Liquid
MDL No.: MFCD00804270

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR