[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride Pd 15,8 --> 16,8 %

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: [1,1'-Bis(di-tert-butylphosphino)ferrocene]dichloropalladium(II), 1,1'-Bis(di-tert-butylphosphino)ferrocene-palladium dichloride, Dichloro[1,1'-bis(di-t-butylphosphino)ferrocene]palladium(II), Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)

452940010 452940050 452942500
452940010.EA 205 GBP
452940010. 452940050. 452942500.
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride Pd 15,8 --> 16,8 %
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride
Formula: C₂₆H₄₄Cl₂FeP₂Pd
MW: 651.75 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08064219
CAS Number: 95408-45-0

Order Now

Specification Test Results

Appearance Beige to dark brown Powder
Infrared spectrum Conforms
Titration Argentometric 10.1 to 11.6 % (Cl)
Palladium (Pd) 15.8 to 16.8 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR