Cesium nitrate, ionization buffer solution, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

45294.AEEA 69.8 GBP
45294.AE
Cesium nitrate, ionization buffer solution, Specpure®
Buffers Ionisation Buffers

CAS No.: 7789-18-6
Molecular Formula: 1% Cs (as Cs₂CO₃) in 5% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00010963

Order Now

SPECIFICATIONS

Environmentally Preferable
Physical form Liquid
CAS No. 7789-18-6
MDL No. MFCD00010963
Molecular formula 1% Cs (as Cs2CO3) in 5% HNO3
UN No. 3264

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR