Nickel 61, 61 Ni, plasma standard solution, 61 Ni 10 µg/ml, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

45282.ADEA 274 GBP
45282.AD
Nickel 61, 61 Ni, plasma standard solution, 61 Ni 10 µg/ml, Specpure®
Standards Cationic Standards Nickel Standards

Molecular Formula: Matrix: 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00144901

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No. MFCD00144901

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR