Nickel iron molybdenum, foil, annealed, Thickness: 0.102 mm (0.004 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

45092.HB 45092.4K
45092.HBEA 63.5 GBP
45092.HB 45092.4K
Nickel iron molybdenum, foil, annealed, Thickness: 0.102 mm (0.004 in)
Nickel iron molybdenum

Molecular Formula: Ni:Fe:Mo; 80:15:4.2 wt% Physical Form: ≈40.1g/150mmx300mm MDL No.: MFCD09910537

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR