Palladium 5% on activated carbon wetted with ca. 50% water, powder Type A405023-5

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

45051.06 45051.22 45051.14
45051.06EA 75.2 GBP
45051.06 45051.22 45051.14
Palladium 5% on activated carbon wetted with ca. 50% water, powder Type A405023-5
Palladium

CAS No.: 7440-05-3 Physical Form: Notes: Sold on a dry weight basis Odour: Odourless MDL No.: MFCD00011167

Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD03457879
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR