J.T.Baker®, Microtube Racks
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
45003-0000EA 44.8 GBP
45003-0000 45000-0000 45005-0000 45000-0008
J.T.Baker®, Microtube Racks
Tubes Microtubes
Safe sample storage system.
  • V bottom for optimal sample recovery
  • Individual tubes or strips of eight tubes available
  • 96-well capmat and strips of eight plug caps available
  • Refillable alphanumeric marked rack
  • Non sterile
Microtube racks are supplied with tubes included. Rack can be also filled with individual tubes available separately. They have a footprint in accordance to SBS standard and it is compatible with all current robotic systems.