J.T.Baker® BAKER-flex®, TLC Sheets, Precoated
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4467-02EA 317 GBP
4467-02 4467-04
J.T.Baker® BAKER-flex®, TLC Sheets, Precoated
TLC Plates
Baker-flex® pre-coated flexible polyester sheets have a hard, abrasion resistant coating and solvent resistant substrate for rapid, clean separations. The sheets can be written on and cut to size.
Order Now
SPECIFICATIONS
Environmentally Preferable