Copper etchant

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

44583.WY
44583.WYEA 68.3 GBP
44583.WY
Copper etchant
Copper etchant

Applications: Iron (III) chloride solution for immersion etching copper, copper alloys and Kovar in printed circuit board applications


Physical Form: Liquid Density: 1.19 Appearance: Dark yellow to brown Odour: Acidic UN No.: UN3264 MDL No.: MFCD00161912


Applications: Iron (III) chloride solution for immersion etching copper, copper alloys and Kovar in printed circuit board applications

Density: 1.19
Storage Temperature: Ambient
MDL Number: MFCD00161912
UN: 3264
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR