Zinc oxide 99 Metals basis, powder_nano

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Zinci oxidum, Zinc (II) oxide

44533.09 44533.18
44533.09EA 113 GBP
44533.09 44533.18
Zinc oxide 99 Metals basis, powder_nano
Zinc oxide

Purity: 99% CAS No.: 1314-13-2 Molecular Formula: ZnO Formula Weight: 81.37 Physical Form: 20-30nm APS Powder Density: 5.606 Appearance: White Odour: Odourless Melting Point: 1975° Merck Reference: 14,10147 UN No.: 3077 MDL No.: MFCD00011300

Formula: ZnO
MW: 81.39 g/mol
Density: 5.606
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9 III
REACH: 01-2119463881-32
Merck Index: 14,10147

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR