Melatonin, Millipore®

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide

444300-1
444300-1EA 63.5 GBP
444300-1
Melatonin, Millipore®
Melatonine
Formula: C₁₃H₁₆N₂O₂
MW: 232.28 g/mol
MDL Number: MFCD00005655
CAS Number: 73-31-4
EINECS: 200-797-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR