Anti-MMP9 Rabbit Polyclonal Antibody

Supplier: Merck Millipore (Oncogene)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

444236-500
444236-500EA 453 GBP
444236-500
Anti-MMP9 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MMP9 (matrix metallopeptidase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR