Vortex mixers, large capacity, Multi Vortex-Genie® and Vortex Genie® Max
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SI-M266 SI-M276 SI-MX202 SI-MX203 SI-MX204
444-1293EA 0 GBP
444-1293 SCIISI-M276 SCIISI-MX202 444-1301 SCIISI-MX204
Vortex mixers, large capacity, Multi Vortex-Genie® and Vortex Genie® Max
Shakers Vortex Shakers
Vortex mixers with digital speed control. These units provide accuracy, reproducibility and repeatability with complete control of speed and time. Heavy zinc die casting with specially designed feet prevents 'walking'.
  • Ideal for use with open containers due to programmable gentle ramp up to speed
  • Adjustable clamping systems make these vortex mixers easy to use and allow quick changes from one type of vessel to another
Multi Vortex-Genie® provides large capacity shaking with variable speed control from gentle orbital shaking to more vigorous vortexing.
More Product Information