Platinum 5% on alumina, powder, reduced

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

44222.14 44222.22 44222.06
44222.14EA 378 GBP
44222.14 44222.22 44222.06
Platinum 5% on alumina, powder, reduced
Platinum

CAS No.: 7440-06-4 MDL No.: MFCD03458043

Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR