VWR®, Stirring Rods
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
442-4537EA 35 GBP
442-4537 442-4538 442-4540 442-4543 442-4544 442-4545 442-4546 442-4547 442-4548
VWR®, Stirring Rods
Stirrers Stirring Rods
PTFE with steel core.
  • Inert and will not scratch, can be used up to 280 °C