VWR®, Stirring Rods
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
442-4539EA 24.8 GBP
442-4539 442-4541 442-4542
VWR®, Stirring Rods
Stirrers Stirring Rods
Solid PTFE.
  • Inert and will not scratch, can be used up to 280 °C