Magnetic Stirrers with Remote Control, Multi-Position
Supplier: THERMO H + P LABORTECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
50088111 50088091 50088077 50088078 50088070 50088030 50088034 50088036 50088025 50088009 50088011
442-3323EA 3310 GBP
442-3323 442-3371 442-3372 442-3373 442-3324 442-3381 442-3382 442-3383 442-3222 442-3392 442-3393
Magnetic Stirrers with Remote Control, Multi-Position
Stirrers Magnetic Stirrers, Multi-position
Compact magnetic stirrers with VARIOMAG drive, hermetically sealed in resistant stainless steel housing (except MINI units which are plastic), making these stirrers ideal for use directly in water baths. MINI stirrers are ideal for integration in analytical instruments, cuvette holders or photometers due to their size, however these plastic units are not designed for use directly in baths. There are units with up to 60 stirring positions in this range, and a choice of three external control units. The TELEMODUL 7 has one power setting, the TELEMODUL 20 C has four power settings: 25%; 50%; 75%More Product Information