VWR® Advanced Magnetic Hotplate Stirrers
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
442-1268EA 720 GBP
442-1268 442-1270 442-1265 442-1274
VWR® Advanced Magnetic Hotplate Stirrers
Stirrers Magnetic Hotplate Stirrers
VWR® Advanced magnetic hotplate stirrers are designed to deliver accurate and repeatable results in all research, academic and industrial applications. With temperature ranges up to 500 °C, and stirring speeds reaching 1600 rpm, VWR® magnetic hotplate stirrers provide proper mixing and superior temperature control. Available in three sizes and a choice of two top plate materials.
  • Excellent temperature uniformity and powerful stirring
  • Easy to read, large LCD displays temperatures and speed
  • Industry-leading safety features
  • Cool touch and chemical resistant
More Product Information