Capillary viscometers, Cannon-Fenske
Supplier: SI Analytics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
285402124 285402132 285402149 285402157 285402165 285402173 285402181 285402198 285402202 285402219 285402227
442-1195EA 377 GBP
442-1195 442-1196 442-1197 442-1198 442-1199 442-1217 442-1218 442-1219 442-1228 442-1229 442-1230
Capillary viscometers, Cannon-Fenske
Viscometers Capillary Viscometers
Capillary viscometers calibrated for automatic measurements in routine applications.
Certifications: ISO/DIN 5105, ASTM D 2515, D 446