Multi-position magnetic hotplate stirrers
Supplier: 2MAG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
66306 66306_CH 66306_UK 66315_UK 66315 66315_CH
442-1110EA 5600 GBP
442-1110 442-1111 442-1131 442-0948 442-0946 442-0947
Multi-position magnetic hotplate stirrers
Stirrers Magnetic Hotplate Stirrers, Multi-position
Hotplate stirrers that have 6 or 15 powerful stirring positions. The units have a clear digital display for settings of stirrer speed, stirrer power and temperature of the integrated heater. The SoftStart procedure ensures reliable centring and safe acceleration of the stirring bar. The heating plate is made of aluminium alloy, PTFE coated for chemical resistance and easy cleaning.
  • Maintenance- and wear-free inductive drive
  • Stirrer has 10-step power settings
  • 100% synchronised, jerk-free stirring
  • Independent over-temperature protection inside the heating
More Product Information