Magnetic hotplate stirrers, Undergrad UC152/D and US152/D
Supplier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
UC152D
442-1075EA 0 GBP
442-1075
Magnetic hotplate stirrers, Undergrad UC152/D and US152/D
Stirrers Magnetic Hotplate Stirrers
Compact general purpose hotplate stirrers designed for safety and performance. The hotplate has an innovative LED temperature indicator scale and can also be used in conjunction with the SCT1 digital contact thermometer to accurately control sample temperature. Powerful magnets and motor give stirring speeds up to 2000 min⁻¹ and unit is capable of mixing large volumes (up to 15 litres*). Both models have an integral fitting for a retort rod.
  • Choice of top plate: Robust coated aluminium or chemically resistant ceramic
  • Flashing 'hot' warning light whenever
More Product Information