DrySyn® heating blocks
Supplier: ASYNT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ADS1 ADS2 ADS5 ADS3 ADS4 ADS6 ADS14-50 ADS14
460-0124EA 1100 GBP
460-0124 442-0872 460-0126 460-0125 442-0851 442-0856 ASYNADS14-50 442-1238
DrySyn® heating blocks
Block Heaters Blocks
The DrySyn heating block range is the alternative to oil baths and heating mantles. The systems offer clean, safe synthesis for single or multiple reactions.
  • Compatible with any magnetic hotplate stirrer
  • Safe and rapid temperature ramping to over 300 ºC
  • Accommodates the full range of standard round bottomed flasks, compatible with multi neck flasks
  • Low well design prevents cracking of glassware and increases reaction visibility
  • Chemical and solvent resistant clear anodised finish
  • Heat-resistant handles for safe and easy lifting