Magnetic stirrer, MAXdrive
Supplier: 2MAG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
94400 94400_UK 94400_CH 50001_UK 50001_CH 50001
442-0837EA 1320 GBP
442-0837 442-0839 442-0838 TMAG50001_UK TMAG50001_CH 442-0823
Magnetic stirrer, MAXdrive
Stirrers Magnetic Stirrers
Robust magnetic stirrer with hermetically sealed stainless-steel housing, suitable for highly viscous media. Operating temperatures up to 50 °C.
  • Very strong magnetic coupling by SmCo magnets, for magnetic power transmission over wide distances, for thick vessel bottoms or for double-coat vessels
  • Ideal for integration into process engineering systems
IP protection class: IP 65.
MAXcontrol (external control unit must be ordered separately). Speed range: 100 - 1200 min⁻¹.
Stainless steel control unit for the connection of one stirring driveMore Product Information