Magnetic stirrer, cuvetteMIXdrive 1
Supplier: 2MAG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
40400 90100 90150 90140
442-0812EA 375 GBP
442-0812 442-0767 TMAG90150 TMAG90140
Magnetic stirrer, cuvetteMIXdrive 1
Stirrers Magnetic Stirrers
This miniaturised stirrer for cuvettes is ideal for system integration e.g. in a spectrophotometer. The hermetically sealed stainless steel housing is water, germ and dust-proof. Operating temperatures up to 50 °C. M1.4 thread is integrated at the base.
  • Maintenance- and wear-free inductive drive
  • Jerk-free mixing
  • Submersible in water and oil baths
  • Suitable for use in CO₂ incubators
IP protection class: IP 68.
MIXcontrol eco (external control unit must be ordered separately). Speed range: 120 - 1200 min⁻¹.
Control unit in plastic moulding forMore Product Information