Magnetic hotplate stirrers, C-MAG HS series
Supplier: IKA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9015900 3581200 3581002 3581003 3581202 3581400 3581402 3581403 3581000 3581200 3581202 3581203 9015900 9015902 9015903
IKAA9015900EA 0 GBP
IKAA9015900 442-1103 442-0711 442-0716 IKAA3581202 442-0718 442-0713 442-0719 442-0715 442-0742 442-0712 442-0717 442-0682 442-0743 442-0744
Magnetic hotplate stirrers, C-MAG HS series
Stirrers Magnetic Hotplate Stirrers
Compact stirrers with a chemically resistant ceramic top. The elevated control panel minimises contact with spills and enables easy reading and adjustment of the temperature. Units feature a digital error code display, a 'hot' top indicator to warn user that surface is hot and the strong magnetic adhesion prevents stirring bar decoupling.
  • Microprocessor control sends feedback to the motor and hotplate to maintain constant speed and set temperature
  • C-MAG HS 7 and C-MAG HS 10 can be used with a contact thermometer e.g. the ETS-D5, enabling precise
More Product Information