VWR®, Magnetic stirring bars, rectangular
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
442-0486EA 8.4 GBP
442-0486 442-0487 442-0488 442-0489
VWR®, Magnetic stirring bars, rectangular
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Smooth surface. Very effective stirrers, especially at low speeds.
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)