VWR®, Magnetic Stirring Bars, Octagonal, With Pivot Ring
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
442-0281EA 26.8 GBP
442-0281 442-0282 442-0283 442-0284 442-0286 442-0345 442-0346 442-0347 442-0349 442-0350 442-0352 442-0354 442-0355 442-0356 442-0357 442-0358 442-0423 442-0424 442-0425 442-0426 442-0427 442-0428 442-0429 442-0430 442-0431 442-0432 442-0433 442-0434 442-0435 442-0436 442-0437 442-0438 442-0439 442-0440 442-0441 442-0442 442-0443 442-0444 442-0445 442-0446
VWR®, Magnetic Stirring Bars, Octagonal, With Pivot Ring
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Smooth surface. For use in containers with bases that are slightly uneven or curved.
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Caution: Coloured PTFE coatings are not as inert as pure PTFE.