Magnetic Stirring Bars with Pivot Ring, PTFE
Supplier: COWIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
001.312 001.375.13 SAN001.3108
442-0091EA 4.7 GBP
442-0091 442-0092 COWISAN001.3108
Magnetic Stirring Bars with Pivot Ring, PTFE
Stirrers Magnetic Stirring Bars
PTFE covered, suitable for use in containers with bases that are slightly curved or uneven.
  • Very good chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)