Tungsten ≥99.95% (metals basis), wire, annealed, Ø 0.05 mm (0.002 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

44191.H4 44191.H9
44191.H4EA 71.9 GBP
44191.H4 44191.H9
Tungsten ≥99.95% (metals basis), wire, annealed, Ø 0.05 mm (0.002 in)
Tungsten

Purity: 99.95% CAS No.: 7440-33-7 MDL No.: MFCD00011461

Formula: W
Boiling Pt: 5660 °C (1013 hPa)
Melting Pt: 3410 °C
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR