Overhead stirrer shafts
Supplier: COWIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
005.440.2 005.450.2 005.455.2 005.465.2 005.240.2 005.250.2 005.255.2 005.265.2 005.340.2 005.350.2 005.355.2 005.365.2 005.355.1.2 005.365.1.2 005.0265.12.2 005.0465.12.2 005.255.10.2 005.430.8.2 005.440.8.2 005.450.8.2 005.455.10.2 005.0465.10.2 005.0475.10.2 005.0475.12.2 005.04100.12.2 005.330.8.2 005.340.8.2 005.350.8.2 005.355.10.2 005.0365.10.2 005.0375.10.2 005.0365.12.2 005.0375.12.2 005.03100.12.2 005.230.8.2 005.240.8.2 005.250.8.2 005.0265.10.2 005.0275.10.2 005.0275.12.2 005.02100.12.2 005.0475.16.2 005.0465.2 005.04100.16.2 005.0375.16.2 005.0375.1.2 005.0365.1.2 005.03100.16.2 005.0275.2 005.230.2 005.02100.16.2 005.430.2 005.0275.16.2 005.0475.2 005.330.2 005.0265.2
441-9521EA 50 GBP
441-9521 441-9522 441-9523 441-9524 441-9530 441-9532 441-9533 441-9534 441-9571 441-9572 441-9573 441-9574 441-9575 441-9576 441-0435 COWI005.0465.12.2 441-0449 441-0451 441-0452 441-0453 441-0454 441-0443 441-0445 441-0446 441-0441- 441-0377 441-0378 441-0379 441-0380 441-0366 441-0369 441-0367 441-0370 441-0363 441-0373 441-0374 441-0375 441-0434 441-0437 441-0438 441-0432 441-0447 441-0444 441-0442 441-0371 441-0368 441-0365 441-0364 441-0440 441-0372 441-0433 441-0450 441-0439 441-0448 441-0376 441-0436
Overhead stirrer shafts
Stirrers Stirring Tools
PTFE, with encapsulated steel reinforced drive shaft. Chemically inert. The smooth surface reduces contamination and makes cleaning easier.
  • Non scratch, prevents agitation vessels from being damaged
  • Temperature resistant from −200 to +280 °C
Caution: EX denotes exposed end, these stirrers have a stainless steel shaft core with an exposed end for rigid clamping.
Order Now
SPECIFICATIONS
Environmentally Preferable