Terbium ≥99.9% (metals basis), rods, Ø 12.7 mm (0.50 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

44049.DH 44049.HF
44049.DHEA 1591 GBP
44049.DH 44049.HF
Terbium ≥99.9% (metals basis), rods, Ø 12.7 mm (0.50 in)
Terbium

Purity: 99.9% CAS No.: 7440-27-9 Molecular Formula: Packaged in mylar under argon Physical Form: ≈1.04g/mm, Cast rod, diameter is approximate MDL No.: MFCD00011256

Formula: Tb
Boiling Pt: 3041 °C (1013 hPa)
Melting Pt: 1356 °C
Density: 8.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011256
CAS Number: 7440-27-9
EINECS: 231-137-6
Merck Index: 12,09300

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR