Tungsten ≥99.95% (metals basis), rods 2 mm (0.08 in) Ø

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43971.BV 43971.9X 43971.CE
43971.BVEA 63.1 GBP
43971.BV 43971.9X 43971.CE
Tungsten ≥99.95% (metals basis), rods 2 mm (0.08 in) Ø
Tungsten
Formula: W
Boiling Pt: 5660 °C (1013 hPa)
Melting Pt: 3410 °C
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR