Tetraammineplatinum(II) nitrate (≥50% Pt) ≥99.99% (metals basis), Premion®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

43897.01 43897.03 43897.06
43897.01EA 72.7 GBP
43897.01 43897.03 43897.06
Tetraammineplatinum(II) nitrate (≥50% Pt) ≥99.99% (metals basis), Premion®
Tetraammineplatinum(II) nitrate

Purity: 99.99% CAS No.: 20634-12-2 Molecular Formula: H12N6O6Pt Formula Weight: 387.21 Physical Form: Crystalline Appearance: White Melting Point: 262° dec. UN No.: UN1477 MDL No.: MFCD00011624

Formula: H₁₂N₆O₆Pt
Melting Pt: 262 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011624
CAS Number: 20634-12-2
EINECS: 243-929-9
UN: 1477
ADR: 5.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR