3-Phenyl-2-propyn-1-ol ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43778.09 43778.04 43778.18
43778.09EA 74 GBP
43778.09 43778.04 43778.18
3-Phenyl-2-propyn-1-ol ≥98%
3-Phenyl-2-propyn-1-ol

Purity: 98+% CAS No.: 1504-58-1 Molecular Formula: C6H5C≡CCH2OH Formula Weight: 132.16 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.078 Refractive Index: 1.5867 Boiling Point: 129-130°/10mm Beilstein Reference: 2040911 MDL No.: MFCD00040914

Formula: C₉H₈O
Boiling Pt: 129…130 °C (10 mmHg)
Density: 1.078
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040914
CAS Number: 1504-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR