Propagyl amine hydrochloride ≥95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Propargylamine hydrochloride, Prop-2-yn-1-amine hydrochloride

43775.03 43775.06
43775.03EA 32.8 GBP
43775.03 43775.06
Propagyl amine hydrochloride ≥95%
Propagyl amine hydrochloride

Purity: 95% CAS No.: 15430-52-1 Molecular Formula: HC≡CCH2NH2.HCl Formula Weight: 91.54 Physical Form: Powder Sensitivity: Hygroscopic UN No.: 2811 MDL No.: MFCD00012907

Formula: C₃H₆ClN
MW: 91.54 g/mol
Melting Pt: 179…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00012907
CAS Number: 15430-52-1
EINECS: 239-440-5
UN: 2811
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR