Niobium(V) chloride ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Niobium pentachloride

Packaged under argon.

43729.22 43729.14 43729.06
43729.22EA 736 GBP
43729.22 43729.14 43729.06
Niobium(V) chloride ≥99.999% (metals basis), Puratronic®
Niobium (V) chloride

Purity: 99.999% CAS No.: 10026-12-7 Molecular Formula: NbCl5 Formula Weight: 270.20 Physical Form: Powder, Packaged under argon Density: 2.74 Melting Point: 208.3° Boiling Point: 248.2° Merck Reference: 14,6557 UN No.: 3260 MDL No.: MFCD00011127

Formula: Cl₅Nb
MW: 270.17 g/mol
Boiling Pt: 248.2 °C
Melting Pt: 208.3 °C
Density: 2.74
Storage Temperature: Ambient
MDL Number: MFCD00011127
CAS Number: 10026-12-7
EINECS: 233-059-8
UN: 3260
ADR: 8 III
Merck Index: 14,6557

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR